Learn More
4-Nitrotoluene-d7, TRC
Product Code. 30474573
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474573 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30474573 Supplier Toronto Research Chemicals Supplier No. TRCN57231250MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Nitrotoluene-d7
CAS 84344-19-4
Molecular Formula C7 2H7 N O2
Quantity 50 mg
Synonym Benzene-1,2,4,5-d4','3-(methyl-d3)-6-nitro-, 3-(Methyl-d3)-6-nitrobenzene-1,2,4,5-d4, 4-Nitrotoluene-d7, p-Nitrotoluene-d7, 4-Nitrotoluene D7
InChI Formula InChI=1S/C7H7NO2/c1-6-2-4-7(5-3-6)8(9)10/h2-5H,1H3/i1D3,2D,3D,4D,5D
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1[N+](=O)[O-])C([2H])([2H])[2H]
IUPAC Name 1,2,4,5-tetradeuterio-3-nitro-6-(trideuteriomethyl)benzene
Molecular Weight (g/mol) 144.18
Formula Weight 144.0916