Learn More
4-Nitro-5-desnitro Tinidazole, TRC
Product Code. 30465938
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465938 5 mg 5mg
30482570 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30465938 Supplier Toronto Research Chemicals Supplier No. TRCN4937855MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25459-12-5
Molecular Formula C8 H13 N3 O4 S
Molecular Weight (g/mol) 247.27
Synonym 1-[2-(Ethylsulfonyl)ethyl]-2-methyl-4-nitro-1H-imidazole, 1-[2-(Ethylsulfonyl)ethyl]-2-methyl-4-nitro-imidazole, Tinidazole Imp. B (EP), Tinidazole RC B (USP)
IUPAC Name 1-(2-ethylsulfonylethyl)-2-methyl-4-nitroimidazole
SMILES CCS(=O)(=O)CCn1cc(nc1C)[N+](=O)[O-]
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C8H13N3O4S/c1-3-16(14,15)5-4-10-6-8(11(12)13)9-7(10)2/h6H,3-5H2,1-2H3
Formula Weight 247.0627
Chemical Name or Material 1-[2-(ethylsulfonyl)ethyl]-2-methyl-4-nitro-1H-imidazole