Learn More
4-Nitro-4’-acetylaminodiphenyl Sulfone, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Nitro-4'-acetylaminodiphenyl Sulfone |
| CAS | 1775-37-7 |
| Molecular Formula | C14 H12 N2 O5 S |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C14H12N2O5S/c1-10(17)15-11-2-6-13(7-3-11)22(20,21)14-8-4-12(5-9-14)16(18)19/h2-9H,1H3,(H,15,17) |
| SMILES | CC(=O)Nc1ccc(cc1)S(=O)(=O)c2ccc(cc2)[N+](=O)[O-] |
| IUPAC Name | N-[4-(4-nitrophenyl)sulfonylphenyl]acetamide |
| Molecular Weight (g/mol) | 320.32 |
| Formula Weight | 320.0467 |