Learn More
4-(n-Nonyl-1,1,2,2-d4)-phenol-2,3,5,6-d4, CDN
Product Code. 30313878
Change view
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30313878 0.01 g 0.01g
30311256 0.05 g 0.05g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30313878 Supplier CDN Supplier No. CDND79700.01G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-n-Nonylphenol D8 (ring D4-ethylD4)
CAS 2733716-95-3
Molecular Formula C15 D8 H16 O
Quantity 0.01 g
Synonym 4-n-Nonylphenol D8 (ring D4-ethylD4), 4-Nonylphenol D8 (ring D4 ethyl D4), 4-(n-Nonyl-1,1,2,2-d4)-phenol-2,3,5,6-d4
InChI Formula InChI=1 S/C15H24O/c1-2-3-4-5-6-7-8-9-14-10-12-15(16)13-11-14/h10-13,16 H,2-9H2,1H3/i8D2,9D2,10 D,11 D,12 D,13 D
SMILES [2 H]c1c([2 H])c(c([2 H])c([2 H])c1O)C([2 H])([2 H])C([2 H])([2 H])CCCCCCC
IUPAC Name 2,3,5,6-tetradeuterio-4-(1,1,2,2-tetradeuteriononyl)phenol
Molecular Weight (g/mol) 228.3998
Formula Weight 228.2329 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity