Learn More
4'-(N-Hydroxysulfanilyl)acetanilide, TRC
Specifications
Specifications
| Chemical Name or Material | 4'-(N-Hydroxysulfanilyl)acetanilide |
| CAS | 32604-86-7 |
| Molecular Formula | C14H14N2O4S |
| Quantity | 5 mg |
| Synonym | N-[4-[[4-(Hydroxyamino)phenyl]sulfonyl]phenyl]acetamide |
| InChI Formula | InChI=1S/C14H14N2O4S/c1-10(17)15-11-2-6-13(7-3-11)21(19,20)14-8-4-12(16-18)5-9-14/h2-9,16,18H,1H3,(H,15,17) |
| SMILES | CC(=O)Nc1ccc(cc1)S(=O)(=O)c2ccc(NO)cc2 |
| IUPAC Name | N-[4-[4-(hydroxyamino)phenyl]sulfonylphenyl]acetamide |
| Molecular Weight (g/mol) | 306.34 |
| Formula Weight | 306.0674 |