missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylbenzaldehyde, 97%

Product Code. p-7058395
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323067 1 g 1g
11333067 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323067 Supplier Thermo Scientific Alfa Aesar Supplier No. L13438.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1200-14-2 | C11H14O | 162.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1200-14-2
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00040751
InChI Key ARIREUPIXAKDAY-UHFFFAOYSA-N
Synonym benzaldehyde, 4-butyl, p-butylbenzaldehyde, 4-n-butylbenzaldehyde, butylbenzaldehyde, p-n-butylbenzaldehyde, butylbenzaldehyde;, 4-butylbenzaldehyd, 4-butyl-benzaldehyde, p-butyl-benzaldehyde, h
PubChem CID 70988
IUPAC Name 4-butylbenzaldehyde
SMILES CCCCC1=CC=C(C=O)C=C1

Specifications

Melting Point -43°C
Density 0.97
Boiling Point 96°C to 98°C (5 mmHg)
Refractive Index 1.526
Quantity 1 g
Beilstein 2208267
Sensitivity Air Sensitive
Formula Weight 162.23
Percent Purity 97%
Chemical Name or Material 4-n-Butylbenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.