Learn More
4-n-Butoxy-2,6-difluorobenzoic acid, 97%, Thermo Scientific™
Product Code. 15405867
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15405867 1 g 1g
15415867 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15405867 Supplier Thermo Scientific Alfa Aesar Supplier No. H33078.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
missing translation for 'validMainframeMsg'
Denne vare kan ikke returneres. Se returpolicy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 123843-56-1
Molecular Formula C11H12F2O3
Molecular Weight (g/mol) 230.211
MDL Number MFCD22201018
InChI Key CSCQGLAJIXARHJ-UHFFFAOYSA-N
Synonym 4-n-butoxy-2,6-difluorobenzoic acid
PubChem CID 19010178
IUPAC Name 4-butoxy-2,6-difluorobenzoic acid
SMILES CCCCOC1=CC(=C(C(=C1)F)C(=O)O)F
Tekniske data
Melting Point 112°C to 115°C
Quantity 1 g
Formula Weight 230.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-n-Butoxy-2,6-difluorobenzoic acid
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.