Learn More
4-Morpholineethanesulfonic Acid, TRC
Product Code. 30481763
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481763 50 g 50g
30479928 100 g 100g
30466732 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30481763 Supplier Toronto Research Chemicals Supplier No. TRCM72377550G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4432-31-9
Molecular Formula C6 H13 N O4 S
Molecular Weight (g/mol) 195.24
Synonym 3-(N-Morpholino)ethane Sulfonic Acid, 4-Morpholineethanesulfonic acid, 2-(4-Morpholino)ethanesulfonic acid, 2-(4-Morpholinyl)ethanesulfonic acid, 2-(N-Morpholino)ethanesulfonic acid, 2-Morpholinoethanesulfonic acid, MES, MES (buffering agent), Morpholinoethanesulfonic acid, NSC 157116
IUPAC Name 2-morpholin-4-ylethanesulfonic acid
SMILES OS(=O)(=O)CCN1CCOCC1
Specifications
Quantity 50 g
InChI Formula InChI=1S/C6H13NO4S/c8-12(9,10)6-3-7-1-4-11-5-2-7/h1-6H2,(H,8,9,10)
Formula Weight 195.0565
Chemical Name or Material 3-(N-Morpholino)ethane Sulfonic Acid (MES)