Learn More
Melford 4-Methylumbelliferyl-β-D-cellobiopyranoside
Product Code. 30018505
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018505 500 mg 500mg
30018504 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30018505 Supplier Melford Supplier No. M646000.5
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Fluorescent substrate for β-D-Cellobiopyranoside.
Fluorescent substrate for β-D-Cellobiopyranoside.
4-Methylumbelliferone < 50ppm.
Specifications
Quantity 500 mg