Learn More
Melford 4-Methylumbelliferyl-B-D-Xylopyranoside
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Description
Fluorescent substrate for β-D-Xylosidase.
Fluorescent substrate for β-D-Xylosidase.
4-Methylumbelliferone: <50ppm
Specifications
Specifications
| Quantity | 250 mg |