Learn More
4-Methylumbelliferyl a-D-Mannopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methylumbelliferyl a-D-Mannopyranoside |
| CAS | 28541-83-5 |
| Molecular Formula | C16 H18 O8 |
| Quantity | 100 mg |
| Synonym | 2H-1-Benzopyran-2-one, 7-(α-D-mannopyranosyloxy)-4-methyl-, Mannopyranoside, 4-methyl-2-oxo-2H-1-benzopyran-7-yl, α-D- (8CI), 7-(α-D-Mannopyranosyloxy)-4-methyl-2H-1-benzopyran-2-one, 4-Methylumbelliferyl α-D-mannopyranoside, 4-Methylumbelliferyl α-D-mannoside, 4-Methylumbelliferyl α-mannopyranoside, 4'-Methylumbelliferyl α-D-mannopyranoside |
| InChI Formula | InChI=1S/C16H18O8/c1-7-4-12(18)23-10-5-8(2-3-9(7)10)22-16-15(21)14(20)13(19)11(6-17)24-16/h2-5,11,13-17,19-21H,6H2,1H3/t11-,13-,14+,15+,16+/m1/s1 |
| SMILES | CC1=CC(=O)Oc2cc(O[C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@@H]3O)ccc12 |
| IUPAC Name | 4-methyl-7-[(2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one |
| Molecular Weight (g/mol) | 338.31 |
| Formula Weight | 338.1002 |