Learn More
4-Methylumbelliferyl 2,3,4,6-Tetra-O-acetyl-alpha-D-mannopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methylumbelliferyl 2,3,4,6-Tetra-O-acetyl-alpha-D-mannopyranoside |
| CAS | 28541-71-1 |
| Molecular Formula | C24 H26 O12 |
| Quantity | 100 mg |
| Synonym | 2H-1-Benzopyran-2-one, 4-methyl-7-[(2,3,4,6-tetra-O-acetyl-α-D-mannopyranosyl)oxy]-, Mannopyranoside, 4-methyl-2-oxo-2H-1-benzopyran-7-yl, tetraacetate, α-D- (8CI), 4-Methyl-7-[(2,3,4,6-tetra-O-acetyl-α-D-mannopyranosyl)oxy]-2H-1-benzopyran-2-one, 4-Methylumbelliferyl tetra-O-acetyl-α-D-mannopyranoside, 4-Methylumbelliferyl 2,3,4,6-tetra-O-acetyl-α-D-mannopyranoside |
| InChI Formula | InChI=1S/C24H26O12/c1-11-8-20(29)35-18-9-16(6-7-17(11)18)34-24-23(33-15(5)28)22(32-14(4)27)21(31-13(3)26)19(36-24)10-30-12(2)25/h6-9,19,21-24H,10H2,1-5H3/t19-,21-,22+,23+,24+/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@H](Oc2ccc3C(=CC(=O)Oc3c2)C)[C@@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C |
| IUPAC Name | [(2R,3R,4S,5S,6R)-3,4,5-triacetyloxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 506.46 |
| Formula Weight | 506.1424 |