Learn More
Alfa Aesar™ 4-(Methylthio)thiophenol, 96%
Click to view available options
Quantity:
1 g
5 g
Specifications
Melting Point | 12°C to 16°C |
Boiling Point | 116°C to 117°C (3 mmHg) |
Refractive Index | 1.653 |
Quantity | 1 g |
Beilstein | 1931500 |
Sensitivity | Air Sensitive |
Formula Weight | 156.27 |
Percent Purity | 96% |
Chemical Name or Material | 4-(Methylthio)thiophenol |