missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzyl alcohol, 98%

Product Code. p-7026454
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450617 5 g 5g
11460617 25 g 25g
11470617 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450617 Supplier Thermo Scientific Alfa Aesar Supplier No. B22455.06
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3446-90-0 | C8H10OS | 154.227 g/mol

4-(Methylthio)benzenemethanol is used to prepare (4-substituted benzyl)(trifluoromethyl)pyrazoles and -pyrazolones as potent antihyperglycemic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylthio)benzenemethanol is used to prepare (4-substituted benzyl)(trifluoromethyl)pyrazoles and -pyrazolones as potent antihyperglycemic agents.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3446-90-0
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.227
MDL Number MFCD00009706
InChI Key MTXQKSQYMREAGJ-UHFFFAOYSA-N
Synonym 4-methylthio benzyl alcohol, 4-methylthio phenyl methanol, p-methylthiobenzyl alcohol, 4-methylsulfanyl phenyl methanol, 4-methylthiobenzyl alcohol, 4-methylthio benzylalcohol, p-methylthio benzyl alcohol, 4-methylsulfanylphenyl methanol, 4-hydroxymethyl thioanisole
PubChem CID 592968
IUPAC Name (4-methylsulfanylphenyl)methanol
SMILES CSC1=CC=C(C=C1)CO

Specifications

Melting Point 41°C to 43°C
Boiling Point 108°C to 109°C (0.4 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 154.23
Percent Purity 98%
Chemical Name or Material 4-(Methylthio)benzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.