Learn More
(4-Methylbenzyl)triphenylphosphonium bromide, 98+%
Product Code. 11351327
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11351327 25 g 25g
11341327 5 g 5g
2 options
Product Code. 11351327
Brand: Thermo Scientific Alfa Aesar L01562.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 2378-86-1 | C26H24BrP | 447.36 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2378-86-1
Molecular Formula C26H24BrP
Molecular Weight (g/mol) 447.36
MDL Number MFCD00031580
InChI Key CEEKCHGNKYVTTM-UHFFFAOYSA-M
Synonym 4-methylbenzyl triphenylphosphonium bromide, 4-methylphenyl methyl triphenylphosphanium bromide, pubchem2184, acmc-1cfpa, 4-methylphenyl methyl-triphenyl-phosphanium bromide, 4-methylbenzyl triphenylphosphoniumbromide, 4-methylphenyl methyl-triphenylphosphonium bromide, p-methylbenzyl-triphenylphosphomiumbromide, phosphonium, 4-methylphenyl methyl triphenyl-, bromide 1:1
PubChem CID 11797662
SMILES [Br-].CC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Specifications
Melting Point 268°C to 270°C
Quantity 25 g
Beilstein 3810292
IUPAC Name (4-methylphenyl)methyl-triphenylphosphanium;bromide
Formula Weight 447.36
Percent Purity ≥98%
Chemical Name or Material (4-Methylbenzyl)triphenylphosphonium bromide