missing translation for 'onlineSavingsMsg'
Learn More

(4-Methylbenzyl)triphenylphosphonium bromide, 98+%

Product Code. 11341327
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11341327 5 g 5g
11351327 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11341327

Brand: Thermo Scientific Alfa Aesar L01562.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2378-86-1 | C26H24BrP | 447.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2378-86-1
Molecular Formula C26H24BrP
Molecular Weight (g/mol) 447.36
MDL Number MFCD00031580
InChI Key CEEKCHGNKYVTTM-UHFFFAOYSA-M
Synonym 4-methylbenzyl triphenylphosphonium bromide, 4-methylphenyl methyl triphenylphosphanium bromide, pubchem2184, acmc-1cfpa, 4-methylphenyl methyl-triphenyl-phosphanium bromide, 4-methylbenzyl triphenylphosphoniumbromide, 4-methylphenyl methyl-triphenylphosphonium bromide, p-methylbenzyl-triphenylphosphomiumbromide, phosphonium, 4-methylphenyl methyl triphenyl-, bromide 1:1
PubChem CID 11797662
SMILES [Br-].CC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 268°C to 270°C
Quantity 5 g
Beilstein 3810292
IUPAC Name [(4-methylphenyl)methyl]triphenylphosphanium bromide
Formula Weight 447.36
Percent Purity ≥98%
Chemical Name or Material (4-Methylbenzyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.