missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)pyridine, 99%, Thermo Scientific Chemicals

Catalog Number p-7047684
Click to view available options
Quantity:
1 g
5 g
25 g
1121-58-0
C6H8N2
108.14
MFCD00152625
LSCYTCMNCWMCQE-UHFFFAOYSA-N
4-methylamino pyridine, 4-methylaminopyridine, n-methyl-4-pyridinamine, 4-pyridinamine, n-methyl, pyridin-4-yl methyl amine, 4-mamp, dmap on polystyrene, pubchem12915, pyridin-4yl-methylamine, pyridin-4-yl-methylamine
123098
N-methylpyridin-4-amine
CNC1=CC=NC=C1
1121-58-0
C6H8N2
108.14
MFCD00152625
LSCYTCMNCWMCQE-UHFFFAOYSA-N
4-methylamino pyridine, 4-methylaminopyridine, n-methyl-4-pyridinamine, 4-pyridinamine, n-methyl, pyridin-4-yl methyl amine, 4-mamp, dmap on polystyrene, pubchem12915, pyridin-4yl-methylamine, pyridin-4-yl-methylamine
123098
N-methylpyridin-4-amine
CNC1=CC=NC=C1

4-(Methylamino)pyridine is employed as efficient nucleophilic catalyst during the preparation of ultra-high surface area emulsion templated porous polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylamino)pyridine is employed as efficient nucleophilic catalyst during the preparation of ultra-high surface area emulsion templated porous polymers.

Solubility
Soluble in water, acetone, ethanol, ether and benzene.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 124°C to 128°C
Quantity 1 g
Beilstein 108213
Solubility Information Soluble in water,acetone,ethanol,ether and benzene.
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 4-(Methylamino)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'