Learn More
4-Methyl-5-(2-phosphorylethyl)thiazole, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methyl-5-(2-phosphorylethyl)thiazole |
| CAS | 3269-79-2 |
| Molecular Formula | C6H10NO4PS |
| Quantity | 10 mg |
| Synonym | 5-(β-Hydroxyethyl)-4-methylthiazole monophosphate |
| InChI Formula | InChI=1S/C6H10NO4PS/c1-5-6(13-4-7-5)2-3-11-12(8,9)10/h4H,2-3H2,1H3,(H2,8,9,10) |
| SMILES | CC1=C(SC=N1)CCOP(O)(O)=O |
| Molecular Weight (g/mol) | 223.18 |
| Formula Weight | 223.01 |