missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitropyridine, 98%, Thermo Scientific™

Product Code. p-7044609
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341897 1 g 1g
11351897 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341897 Supplier Thermo Scientific Alfa Aesar Supplier No. L07398.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5832-44-0
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00051829
InChI Key JLNRJMGYBKMDGI-UHFFFAOYSA-N
Synonym 3-nitro-4-picoline, 4-methyl-3-nitro-pyridine, 3-nitro-4-methylpyridine, pyridine, 4-methyl-3-nitro, zlchem 957, pubchem6644, 4-methyl-3-nitro pyridine, acmc-1b13v, ksc490s2l, tpc-py011
PubChem CID 521828
IUPAC Name 4-methyl-3-nitropyridine
SMILES CC1=C(C=NC=C1)[N+](=O)[O-]

Specifications

Melting Point 18°C to 22°C
Density 1.23
Boiling Point 238°C
Flash Point 108°C (226°F)
Refractive Index 1.546
Quantity 1 g
UN Number UN2810
Beilstein 118796
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 4-Methyl-3-nitropyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.