Learn More
4-Methyl-2-oxovaleric Acid, TRC
Product Code. 30010809
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010809 100 mg 100mg
30012575 50 mg 50mg
30012003 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010809 Supplier Toronto Research Chemicals Supplier No. TRCM220748100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 816-66-0
Molecular Formula C6 H10 O3
Molecular Weight (g/mol) 130.14
Synonym 4-Methyl-2-oxovaleric acid, 2-Oxoisocaproic acid, Isobutylglyoxylic acid, Ketoleucine
IUPAC Name 4-methyl-2-oxopentanoic acid
SMILES CC(C)CC(=O)C(=O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C6H10O3/c1-4(2)3-5(7)6(8)9/h4H,3H2,1-2H3,(H,8,9)
Formula Weight 130.063
Percent Purity >95%
Chemical Name or Material alpha-Ketoisocaproic acid
Grade HPLC