Learn More
4-Methyl-2-(4-piperidinyl)pyridine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methyl-2-(4-piperidinyl)pyridine Hydrochloride |
| CAS | 2770359-55-0 |
| Molecular Formula | C11H18Cl2N2 |
| Quantity | 25 mg |
| Synonym | 4-methyl-2-(piperidin-4-yl)pyridine dihydrochloride |
| InChI Formula | InChI=1S/C11H16N2.2ClH/c1-9-2-7-13-11(8-9)10-3-5-12-6-4-10;;/h2,7-8,10,12H,3-6H2,1H3;2*1H |
| SMILES | CC1=CC(C2CCNCC2)=NC=C1.Cl.Cl |
| IUPAC Name | 4-methyl-2-(piperidin-4-yl)pyridine dihydrochloride |
| Molecular Weight (g/mol) | 249.18 |
| Formula Weight | 248.08 |