Learn More
4-Methyl-2-(4-piperidinyl)pyridine Hydrochloride, TRC
Product Code. 30472256
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472256 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30472256 Supplier Toronto Research Chemicals Supplier No. TRCM256031250MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Methyl-2-(4-piperidinyl)pyridine Hydrochloride
CAS 2770359-55-0
Molecular Formula C11H18Cl2N2
Quantity 250 mg
Synonym 4-methyl-2-(piperidin-4-yl)pyridine dihydrochloride
InChI Formula InChI=1S/C11H16N2.2ClH/c1-9-2-7-13-11(8-9)10-3-5-12-6-4-10;;/h2,7-8,10,12H,3-6H2,1H3;2*1H
SMILES CC1=CC(C2CCNCC2)=NC=C1.Cl.Cl
IUPAC Name 4-methyl-2-(piperidin-4-yl)pyridine dihydrochloride
Molecular Weight (g/mol) 249.18
Formula Weight 248.08