Learn More
4-(Methoxycarbonyl)cubanecarboxylic acid, 97%, ACROS Organics™
Product Code. 11935041 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11935041 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11935041 Supplier Thermo Scientific Acros Supplier No. 444830010
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 24539-28-4
Assay Percent Range 97%
Molecular Formula C11H10O4
Quantity 1 g
Synonym 4-methoxycarbonylcubanecarboxylic acid, 8-methoxycarbonyl cubane-1-carboxylic acid, pentacyclo 4.2.0.02,5.03,8.04,7 octane-1,4-dicarboxylicacid, 1-methyl ester, 1-carboxy-4-carbomethoxycubane, 4-methoxycarbonyl pentacyclo 4.2.0.02,5.03,8.04,7 octane-1-carboxylic acid, 4-methoxycarbonyl-1-cubanecarboxylic acid, cubane-1,4-dicarboxylic acid 1-methyl ester, 4-methoxycarbonyl cubane-1-carboxylic acid, pentacyclo 4.2.0.02,5.03,8.04,7 octane-1,4-dicarboxylic acid, 1-methyl ester
InChI Key ARRJEFLYRAVFJA-UHFFFAOYSA-N
SMILES COC(=O)C12C3C4C1C5C2C3C45C(=O)O
IUPAC Name 4-methoxycarbonylcubane-1-carboxylic acid
Molecular Weight (g/mol) 206.197
PubChem CID 369179
Formula Weight 206.2
Chemical Name or Material 4-(Methoxycarbonyl)cubanecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Methoxycarbonyl)cubanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.