Learn More
4-Methoxybenzaldehyde, MedChemExpress
Product Code. 16489416
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16489416 Supplier Medchem Express Supplier No. HYY0740
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
MedChemExpress 4-Methoxybenzaldehyde is a naturally occurring fragrant phenolic compound. 4-Methoxybenzaldehyde has been found in many plant species including horseradish, anise, star anise. 4-Methoxybenzaldehyde is a possible neurotoxicant and it has shown effects that include mortality, attractancy, and interference with host seeking .
4-Methoxybenzaldehyde is a naturally occurring fragrant phenolic compound. 4-Methoxybenzaldehyde has been found in many plant species including horseradish, anise, star anise. 4-Methoxybenzaldehyde is a possible neurotoxicant and it has shown effects that include mortality, attractancy, and interference with host seeking .
Specifications
Chemical Name or Material 4-Methoxybenzaldehyde
CAS 123-11-5
Color Colorless
Purity Grade Notes Research
Molecular Formula C8H8O2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (734.48 mM; Need ultrasonic)
SMILES O=CC1=CC=C(OC)C=C1
Molecular Weight (g/mol) 136.15
Formula Weight 136.15
Percent Purity 99.67%
Grade Research
Physical Form Liquid
For Use With (Application) Metabolism-protein/nucleotide metabolism