missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-N-(2-methoxybenzyl)benzamide, 97%, Thermo Scientific™

Product Code. 15445298
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445298 1 g 1g
15455298 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15445298 Fornitore Thermo Scientific Alfa Aesar N. del fornitore H59096.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 331440-04-1
Molecular Formula C16H17NO3
Molecular Weight (g/mol) 271.316
MDL Number MFCD00752486
InChI Key OCHBASPTHVVGMS-UHFFFAOYSA-N
Synonym 4-methoxy-n-2-methoxybenzyl benzamide, 4-methoxy-n-2-methoxyphenyl methyl benzamide, cambridge id 5337527, n-2-methoxybenzyl-4-methoxybenzamide, 4-methoxyphenyl-n-2-methoxyphenyl methyl carboxamide
PubChem CID 796001
IUPAC Name 4-methoxy-N-[(2-methoxyphenyl)methyl]benzamide
SMILES COC1=CC=C(C=C1)C(=O)NCC2=CC=CC=C2OC

Specifica

Quantity 1 g
Beilstein 23598862
Formula Weight 271.32
Percent Purity 97%
Chemical Name or Material 4-Methoxy-N-(2-methoxybenzyl)benzamide
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.