Learn More
4-Methoxy-d3-benzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Methoxybenzoic Acid D3 (methoxy-D3) |
| CAS | 27914-54-1 |
| Molecular Formula | C8H5D3O3 |
| Quantity | 50 mg |
| Synonym | p-Anisic-α,α,α-d3 Acid, 4-(Trideuteromethoxy)benzoic Acid |
| InChI Formula | InChI=1S/C8H8O3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3,(H,9,10)/i1D3 |
| SMILES | O=C(O)C1=CC=C(OC([2H])([2H])[2H])C=C1 |
| IUPAC Name | 4-(methoxy-d3)benzoic acid |
| Molecular Weight (g/mol) | 155.17 |
| Formula Weight | 155.07 |