Learn More
4-Methoxy-d3-benzoic Acid, CDN
Product Code. 30324334
Change view
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324334 0.5 g 0.50g
30311391 0.25 g 0.25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30324334 Supplier CDN Supplier No. CDND62310.5G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Methoxybenzoic Acid D3 (methoxy-D3)
CAS 27914-54-1
Molecular Formula C8H5D3O3
Quantity 0.5 g
Synonym p-Anisic-α,α,α-d3 Acid, 4-(Trideuteromethoxy)benzoic Acid
InChI Formula InChI=1 S/C8H8O3/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5 H,1H3,(H,9,10)/i1D3
SMILES O=C(O)C1=CC=C(OC([2 H])([2 H])[2 H])C=C1
IUPAC Name 4-(methoxy-d3)benzoic acid
Molecular Weight (g/mol) 155.17
Formula Weight 155.07 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity