missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. p-7029167
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382459 1 g 1g
11392459 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382459 Supplier Thermo Scientific Alfa Aesar Supplier No. H26606.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
As organic intermediate, pharmaceutical intermediate. In chemical synthesis.

Solubility
Sparingly Soluble (0.16 g/L) (25°C).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261951-89-7
Molecular Formula C9H8BrF3O
Molecular Weight (g/mol) 269.061
MDL Number MFCD01631578
InChI Key FZICFAROPYFJMF-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl benzyl bromide, 4-bromomethyl-1-methoxy-2-trifluoromethyl benzene, benzene, 4-bromomethyl-1-methoxy-2-trifluoromethyl, 4-bromomethyl-1-methoxy-2-trifluoromethyl-benzene, 4-bromomethyl-2-trifluoromethyl-1-methoxybenzene, acmc-20amfh, pubchem4655, 4-methoxy-3-trifluoromethyl benzylbromide
PubChem CID 2775302
IUPAC Name 4-(bromomethyl)-1-methoxy-2-(trifluoromethyl)benzene
SMILES COC1=C(C=C(C=C1)CBr)C(F)(F)F

Specifications

Melting Point 75°C to 79°C
Quantity 1 g
UN Number UN3261
Solubility Information Sparingly Soluble (0.16g/L) (25°C).
Formula Weight 269.06
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.