Learn More
4-Iodopyrimidine, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Iodopyrimidine |
| CAS | 31462-57-4 |
| Molecular Formula | C4H3IN2 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C4H3IN2/c5-4-1-2-6-3-7-4/h1-3H |
| SMILES | IC1=CC=NC=N1 |
| IUPAC Name | 4-iodopyrimidine |
| Molecular Weight (g/mol) | 205.98 |
| Formula Weight | 205.93 |