missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3-nitrobenzoic acid, 97%

Product Code. p-7028357
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491647 5 g 5g
11401657 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491647 Supplier Thermo Scientific Alfa Aesar Supplier No. B25626.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 35674-27-2 | C7H4INO4 | 293.02 g/mol

4-Iodo-3-nitrobenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-3-nitrobenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from light, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35674-27-2
Molecular Formula C7H4INO4
Molecular Weight (g/mol) 293.02
MDL Number MFCD00502399
InChI Key DNMTZLCNLAIKQC-UHFFFAOYSA-N
Synonym 4-iodo-3-nitrobenzoic acid, 4-iodo-3-nitrobenzoicacid, 3-nitro-4-iodobenzoic acid, 4-iodo-3-nitro-benzoic acid, benzoic acid, 4-iodo-3-nitro, pubchem13471, acmc-209z8y, 4-iodo-3-nitro benzoic acid
PubChem CID 11208598
IUPAC Name 4-iodo-3-nitrobenzoic acid
SMILES OC(=O)C1=CC(=C(I)C=C1)[N+]([O-])=O

Specifications

Melting Point 208°C to 211°C
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 293.02
Percent Purity 97%
Chemical Name or Material 4-Iodo-3-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.