Learn More
4-Iodo-3-methylbenzoic Acid Methyl Ester, TRC
Product Code. 30394987
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30394987 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30394987 Supplier Toronto Research Chemicals Supplier No. TRCI7086001G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Iodo-3-methylbenzoic Acid Methyl Ester
CAS 5471-81-8
Molecular Formula C9H9IO2
Quantity 1 g
Synonym 4-Iodo-m-toluic Acid Methyl Ester, Methyl 3-Methyl-4-iodobenzoate, Methyl 4-Iodo-3-methylbenzoate, NSC 28460
InChI Formula InChI=1S/C9H9IO2/c1-6-5-7(9(11)12-2)3-4-8(6)10/h3-5H,1-2H3
SMILES COC(=O)c1ccc(I)c(C)c1
IUPAC Name methyl 4-iodo-3-methylbenzoate
Molecular Weight (g/mol) 276.07
Formula Weight 275.9647