Learn More
4-Iodo-3-methylbenzoic Acid Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Iodo-3-methylbenzoic Acid Methyl Ester |
| CAS | 5471-81-8 |
| Molecular Formula | C9H9IO2 |
| Quantity | 100 mg |
| Synonym | 4-Iodo-m-toluic Acid Methyl Ester, Methyl 3-Methyl-4-iodobenzoate, Methyl 4-Iodo-3-methylbenzoate, NSC 28460 |
| InChI Formula | InChI=1S/C9H9IO2/c1-6-5-7(9(11)12-2)3-4-8(6)10/h3-5H,1-2H3 |
| SMILES | COC(=O)c1ccc(I)c(C)c1 |
| IUPAC Name | methyl 4-iodo-3-methylbenzoate |
| Molecular Weight (g/mol) | 276.07 |
| Formula Weight | 275.9647 |