Learn More
4-Hydroxybenzaldehyde-d4, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxybenzaldehyde-d4 |
| CAS | 284474-52-8 |
| Molecular Formula | C7 D4 H2 O2 |
| Quantity | 100 mg |
| Synonym | Benzaldehyde-2,3,5,6-d4, 4-hydroxy- (9CI, ACI), 4-Hydroxybenzaldehyde-2,3,5,6-d4 (ACI), 4-Hydroxybenzaldehyde-d4, 4-Hydroxybenzaldehyde-2,3,5,6-D4, 4-Formylphenol-2,3,5,6-d4 |
| InChI Formula | InChI=1S/C7H6O2/c8-5-6-1-3-7(9)4-2-6/h1-5,9H/i1D,2D,3D,4D |
| SMILES | [2H]c1c([2H])c(C=O)c([2H])c([2H])c1O |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-hydroxybenzaldehyde |
| Molecular Weight (g/mol) | 126.146 |
| Formula Weight | 126.062 |