Learn More
4-Hydroxybenzaldehyde-2,3,5,6-d4, CDN
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxybenzaldehyde-d4 |
| CAS | 284474-52-8 |
| Molecular Formula | C7 D4 H2 O2 |
| Quantity | 0.1 g |
| Synonym | Benzaldehyde-2,3,5,6-d4, 4-hydroxy- (9 CI, ACI), 4-Hydroxybenzaldehyde-2,3,5,6-d4 (ACI), 4-Hydroxybenzaldehyde-d4, 4-Hydroxybenzaldehyde-2,3,5,6-D4, 4-Formylphenol-2,3,5,6-d4 |
| InChI Formula | InChI=1 S/C7H6O2/c8-5-6-1-3-7(9)4-2-6/h1-5,9 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(C=O)c([2 H])c([2 H])c1O |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-hydroxybenzaldehyde |
| Molecular Weight (g/mol) | 126.146 |
| Formula Weight | 126.062 g/mol |
| Show More |