Learn More
4-Hydroxy-3-methoxy-d3-benzoic Acid, CDN
Specifications
Specifications
| Chemical Name or Material | Vanillic-d3 Acid |
| CAS | 2733717-19-4 |
| Molecular Formula | C8 D3 H5 O4 |
| Quantity | 0.005 g |
| Synonym | Benzoic acid, 4-hydroxy-3-(methoxy-d3)- (ACI), 4-Hydroxy-3-methoxy-d3-benzoic Acid, 4-Hydroxy-3-(methoxy-d3)benzoic Acid, Vanillic-d3 Acid, Vanillic acid-D3 (methoxy-D3) |
| InChI Formula | InChI=1 S/C8H8O4/c1-12-7-4-5(8(10)11)2-3-6(7)9/h2-4,9 H,1H3,(H,10,11)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1cc(ccc1O)C(=O)O |
| IUPAC Name | 4-hydroxy-3-(trideuteriomethoxy)benzoic acid |
| Molecular Weight (g/mol) | 171.1652 |
| Formula Weight | 171.0611 g/mol |
| Show More |