Learn More
4-Hydroxy-3-(2-methyl-1,3-diphenylpentyl)-2H-chromen-2-one, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxy-3-(2-methyl-1,3-diphenylpentyl)-2H-chromen-2-one |
| CAS | 2815450-82-7 |
| Molecular Formula | C27H26O3 |
| Quantity | 10 mg |
| Synonym | 4-Hydroxy-3-(2-methyl-1,3-diphenylpentyl)-2H-1-benzopyran-2-one |
| InChI Formula | InChI=1S/C27H26O3/c1-3-21(19-12-6-4-7-13-19)18(2)24(20-14-8-5-9-15-20)25-26(28)22-16-10-11-17-23(22)30-27(25)29/h4-18,21,24,28H,3H2,1-2H3 |
| SMILES | O=C1C(C(C2=CC=CC=C2)C(C(C3=CC=CC=C3)CC)C)=C(O)C4=CC=CC=C4O1 |
| IUPAC Name | 4-hydroxy-3-(2-methyl-1,3-diphenylpentyl)-2H-chromen-2-one |
| Molecular Weight (g/mol) | 398.49 |
| Formula Weight | 398.19 |