Learn More
4-Hydroxy-13-cis-retinoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxy-13-cis-retinoic Acid |
| CAS | 75281-25-3 |
| Molecular Formula | C20 H28 O3 |
| Quantity | 50 mg |
| Synonym | Retinoic acid, 4-hydroxy-, 13-cis-, 13-cis-4-Hydroxyretinoic acid, 4-Hydroxy-13-cis-retinoic acid, 4-Hydroxyisotretinoin, (2Z,4E,6E,8E)-9-[(3RS)-3-Hydroxy-2,6,6-trimethylcyclohex-1-en-1-yl]-3,7-dimethylnona-2,4,6,8-tetraenoic Acid |
| InChI Formula | InChI=1S/C20H28O3/c1-14(7-6-8-15(2)13-19(22)23)9-10-17-16(3)18(21)11-12-20(17,4)5/h6-10,13,18,21H,11-12H2,1-5H3,(H,22,23)/b8-6+,10-9+,14-7+,15-13- |
| SMILES | C\C(=C\C(=O)O)\C=C\C=C(/C)\C=C\C1=C(C)C(O)CCC1(C)C |
| IUPAC Name | (2Z,4E,6E,8E)-9-(3-hydroxy-2,6,6-trimethylcyclohexen-1-yl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid |
| Molecular Weight (g/mol) | 316.43 |
| Formula Weight | 316.2038 |