Learn More
4-Hydroxy[1,3]dioxolo[4,5-g]cinnoline-3-carbonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxy[1,3]dioxolo[4,5-g]cinnoline-3-carbonitrile |
| CAS | 28657-78-5 |
| Molecular Formula | C10H5N3O3 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C10H5N3O3/c11-3-7-10(14)5-1-8-9(16-4-15-8)2-6(5)12-13-7/h1-2H,4H2,(H,12,14) |
| SMILES | Oc1c(nnc2cc3OCOc3cc12)C#N |
| IUPAC Name | 4-hydroxy-[1,3]dioxolo[4,5-g]cinnoline-3-carbonitrile |
| Molecular Weight (g/mol) | 215.16 |
| Formula Weight | 215.0331 |