Learn More
4-Fmoc-3(S)-morpholinecarboxylic Acid, TRC
Product Code. 30398542
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398542 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30398542 Supplier Toronto Research Chemicals Supplier No. TRCF648485100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Fmoc-3(S)-morpholinecarboxylic Acid
CAS 281655-37-6
Molecular Formula C20 H19 N O5
Quantity 100 mg
Synonym (S)-Fmoc-3-carboxymorpholine, (S)-4-N-Fmoc-3-morpholinecarboxylic Acid, (S)-4-(((9H-Fluoren-9-yl)methoxy)carbonyl) morpholine-3-carboxylic Acid
InChI Formula InChI=1S/C20H19NO5/c22-19(23)18-12-25-10-9-21(18)20(24)26-11-17-15-7-3-1-5-13(15)14-6-2-4-8-16(14)17/h1-8,17-18H,9-12H2,(H,22,23)/t18-/m0/s1
SMILES OC(=O)[C@@H]1COCCN1C(=O)OCC2c3ccccc3c4ccccc24
IUPAC Name (3S)-4-(9H-fluoren-9-ylmethoxycarbonyl)morpholine-3-carboxylic acid
Molecular Weight (g/mol) 353.37
Formula Weight 353.1263