Learn More
4-Fmoc-3(S)-morpholinecarboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Fmoc-3(S)-morpholinecarboxylic Acid |
| CAS | 281655-37-6 |
| Molecular Formula | C20 H19 N O5 |
| Quantity | 500 mg |
| Synonym | (S)-Fmoc-3-carboxymorpholine, (S)-4-N-Fmoc-3-morpholinecarboxylic Acid, (S)-4-(((9H-Fluoren-9-yl)methoxy)carbonyl) morpholine-3-carboxylic Acid |
| InChI Formula | InChI=1S/C20H19NO5/c22-19(23)18-12-25-10-9-21(18)20(24)26-11-17-15-7-3-1-5-13(15)14-6-2-4-8-16(14)17/h1-8,17-18H,9-12H2,(H,22,23)/t18-/m0/s1 |
| SMILES | OC(=O)[C@@H]1COCCN1C(=O)OCC2c3ccccc3c4ccccc24 |
| IUPAC Name | (3S)-4-(9H-fluoren-9-ylmethoxycarbonyl)morpholine-3-carboxylic acid |
| Molecular Weight (g/mol) | 353.37 |
| Formula Weight | 353.1263 |