Learn More
4-Fluorobenzyl-2,3,5,6-d4 Chloride, CDN
Specifications
Specifications
| Chemical Name or Material | 4-Fluorobenzyl Chloride-2,3,5,6-D4 |
| CAS | 1219804-10-0 |
| Molecular Formula | C7 D4 H2 Cl F |
| Quantity | 0.5 g |
| Synonym | 4-Fluorobenzyl-2,3,5,6-d4 Chloride, 4-Fluorobenzyl Chloride-d4, 4-Fluorobenzyl Chloride-2,3,5,6-D4, 1-(Chloromethyl)-4-fluorobenzene-2,3,5,6-d4, α-Chloro-4-fluorotoluene-2,3,5,6-d4 |
| InChI Formula | InChI=1 S/C7H6ClF/c8-5-6-1-3-7(9)4-2-6/h1-4 H,5H2/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(CCl)c([2 H])c([2 H])c1F |
| IUPAC Name | 1-(chloromethyl)-2,3,5,6-tetradeuterio-4-fluorobenzene |
| Molecular Weight (g/mol) | 148.5986 |
| Formula Weight | 148.0393 g/mol |
| Show More |