Learn More
4-Fluorobenzhydrazide, 97%
Product Code. p-7035769
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490378 5 g 5g
15400388 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15490378 Supplier Thermo Scientific Alfa Aesar Supplier No. H53496.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 456-06-4 | C7H7FN2O | 154.144 g/mol
Used in the stnthesis of fluorinated poly (1, 3, 4-oxadiazole-ether-imide). N-(3,5-dinitrobenzoyl)-N?-(4-fluorobenzoyl)-hydrazine with 4-Fluorobenzhydrazide as solvent in NMP.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in the stnthesis of fluorinated poly (1, 3, 4-oxadiazole-ether-imide). N-(3,5-dinitrobenzoyl)-N′-(4-fluorobenzoyl)-hydrazine with 4-Fluorobenzhydrazide as solvent in NMP.
Solubility
Soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 456-06-4
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.144
MDL Number MFCD00060562
InChI Key UIVXXFYJRYVRKJ-UHFFFAOYSA-N
Synonym 4-fluorobenzhydrazide, 4-fluorobenzoic hydrazide, p-fluorobenzoic acid hydrazide, p-fluorobenzoylhydrazine, 4-fluorobenzoic acid hydrazide, hydrazine, p-fluorobenzoyl, 4-fluoro benzoyl hydrazine, benzoic acid, p-fluoro-, hydrazide, 4-fluorobenzenecarbohydrazide, 4-fluoro-benzoic acid hydrazide
PubChem CID 9972
IUPAC Name 4-fluorobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)F
Specifications
Melting Point 160°C to 162°C
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material 4-Fluorobenzhydrazide