missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methoxycarbonylphenylboronic acid, 98%, Thermo Scientific™

Product Code. 15515176 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15515176 1 g 1g
15525176 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15515176 Supplier Thermo Scientific Acros Supplier No. 455880010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 874219-35-9
Molecular Formula C8H8BFO4
Molecular Weight (g/mol) 197.96
InChI Key GQBWFAIHGHHNMM-UHFFFAOYSA-N
Synonym 4-fluoro-3-methoxycarbonyl phenylboronic acid, 4-fluoro-3-methoxycarbonylphenyl boronic acid, 4-fluoro-3-methoxycarbonyl phenyl boronic acid, acmc-209qkz, ksc495k9r, 3-methoxycarbonyl-4-fluorophenylboronic acid, 4-fluoro-3-methoxycarbonylphenyl boronicacid, 4-fluoro-3-methoxycarbonyl-phenyl boronic acid, 4-fluoranyl-3-methoxycarbonyl-phenyl boronic acid
PubChem CID 23005332
IUPAC Name (4-fluoro-3-methoxycarbonylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)F)C(=O)OC)(O)O

Specifications

Melting Point 253.0°C to 255.5°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 197.96
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-methoxycarbonylphenylboronic acid
compliance-icons
Product Identifier
  • 4-Fluoro-3-methoxycarbonylphenylboronic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.