Learn More
4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid, 98%, Thermo Scientific™
Product Code. 15410406
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15410406
Brand: Thermo Scientific Alfa Aesar H53123.MD
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 874219-35-9
Molecular Formula C8H8BFO4
Molecular Weight (g/mol) 197.956
MDL Number MFCD06801688
InChI Key GQBWFAIHGHHNMM-UHFFFAOYSA-N
Synonym 4-fluoro-3-methoxycarbonyl phenylboronic acid, 4-fluoro-3-methoxycarbonylphenyl boronic acid, 4-fluoro-3-methoxycarbonyl phenyl boronic acid, acmc-209qkz, ksc495k9r, 3-methoxycarbonyl-4-fluorophenylboronic acid, 4-fluoro-3-methoxycarbonylphenyl boronicacid, 4-fluoro-3-methoxycarbonyl-phenyl boronic acid, 4-fluoranyl-3-methoxycarbonyl-phenyl boronic acid
PubChem CID 23005332
IUPAC Name (4-fluoro-3-methoxycarbonylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)F)C(=O)OC)(O)O
Specifications
Melting Point 253°C to 255°C
Quantity 250 mg
Formula Weight 197.96
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid