Learn More
4-Fluoro-2-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™
Product Code. 15437605
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437605 250 mg 250mg
11332529 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15437605 Supplier Thermo Scientific Alfa Aesar Supplier No. H26637.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 176225-09-5
Molecular Formula C7H3ClF4O2S
Molecular Weight (g/mol) 262.603
MDL Number MFCD01091000
InChI Key IGMYEVQPXWKFQF-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl benzenesulfonyl chloride, 4-fluoro-2-trifluoromethyl benzene-1-sulfonyl chloride, 4-fluoro-2-trifluoromethyl-benzenesulfonyl chloride, 4-fluoro-2-trifluoromethyl benzenesulphonyl chloride, 2-chlorosulphonyl-4-fluorobenzotrifluoride, 4-fluoro-2-trifluoromethyl-benzenesulfonylchloride, 4-fluoro-2-trifluoromethyl benzene sulphonyl chloride, pubchem4276, acmc-1c2ls, 2-chlorosulphonyl-5-fluorobenzotrifluoride
PubChem CID 2737548
IUPAC Name 4-fluoro-2-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=C(C=C1F)C(F)(F)F)S(=O)(=O)Cl
Specifications
Melting Point 48°C to 51°C
Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 262.61
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzenesulfonyl chloride