Learn More
4-(Ethylsulfurate Sulfonyl)aniline, TRC
Product Code. 30382399
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382399 100 mg
30387356 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30382399 Supplier Toronto Research Chemicals Supplier No. TRCE926000100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2494-89-5
Molecular Formula C8H11NO6S2
Molecular Weight (g/mol) 281.31
Synonym 1-Aminobenzene-4-(2-sulfatoethyl)sulfone, 1-Aminobenzene-4-beta-sulfatoethylsulfone, 2-(4-Aminobenzenesulfonyl)ethyl sulfate, 2-(4-Aminophenylsulfonyl)ethyl bisulfate, 2-[(4-Aminophenyl)sulfonyl]ethyl hydrogen sulfate, 2-[(p-Aminophenyl)sulfonyl]ethyl hydrogen sulfate, 4-(2-Sulfatoethylsulfonyl)aniline, 4-(beta-Sulfatoethylsulfonyl)aniline, 4-Amino-1-beta-sulfatoethylsulfonylbenzene, 4-Aminophenyl 2-sulfatoethyl sulfone, 4-Aminophenyl beta-hydroxyethyl sulfone sulfate ester, 4-Aminophenyl beta-sulfatoethyl sulfone, 4-[(beta-Sulfooxyethyl)sulfonyl]aniline, Para Base Sulfate, Sodium 4-(beta-sulphatoethyl)sulfonylaniline, [2-[(4-Aminobenzene)sulfonyl]ethoxy]sulfonic acid, p-(2-Sulfatoethylsulfonyl)aniline, p-(beta-Sulfatoethylsulfonyl)aniline, p-Aminophenyl 2-sulfatoethyl sulfone, p-Aminophenyl beta-sulfatoethyl sulfone
SMILES Nc1ccc(cc1)S(=O)(=O)CCOS(=O)(=O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C8H11NO6S2/c9-7-1-3-8(4-2-7)16(10,11)6-5-15-17(12,13)14/h1-4H,5-6,9H2,(H,12,13,14)
Formula Weight 281.0028
Chemical Name or Material 4-(Ethylsulfurate Sulfonyl)aniline