Learn More
4-Ethyl-5-fluoropyrimidine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Ethyl-5-fluoropyrimidine Hydrochloride |
| CAS | 1391052-89-3 |
| Molecular Formula | C6H8ClFN2 |
| Quantity | 250 mg |
| Synonym | Voriconazole EP Impurity C HCl |
| InChI Formula | InChI=1S/C6H7FN2.ClH/c1-2-6-5(7)3-8-4-9-6;/h3-4H,2H2,1H3;1H |
| SMILES | Cl.CCc1ncncc1F |
| IUPAC Name | 4-ethyl-5-fluoropyrimidine;hydrochloride |
| Molecular Weight (g/mol) | 162.59 |
| Formula Weight | 162.036 |