missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3,5-difluorobenzyl bromide, 97%, Thermo Scientific™

Product Code. p-7031651
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425567 250 mg 250mg
15415567 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425567 Supplier Thermo Scientific Alfa Aesar Supplier No. H32801.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017779-38-2
Molecular Formula C9H9BrF2O
Molecular Weight (g/mol) 251.071
MDL Number MFCD09258691
InChI Key SFMOQKHETBGLAR-UHFFFAOYSA-N
Synonym 4-ethoxy-3,5-difluorobenzyl bromide, 5-bromomethyl-2-ethoxy-1,3-difluorobenzene, 4-ethoxy-3,5-difluorobenzyl bromide, jrd
PubChem CID 46737611
IUPAC Name 5-(bromomethyl)-2-ethoxy-1,3-difluorobenzene
SMILES CCOC1=C(C=C(C=C1F)CBr)F

Specifications

Refractive Index 1.5183
Quantity 250 mg
UN Number UN3265
Formula Weight 251.07
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-3,5-difluorobenzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.