Learn More
4-Desulfo-4-hydroxy Picosulfate Monosodium Salt, TRC
Product Code. 17914575
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17914575 500 mg 500mg
17991325 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17914575 Supplier Toronto Research Chemicals Supplier No. TRCD297930500MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32500-19-9
Molecular Formula C18 H14 N O5 S . Na
Molecular Weight (g/mol) 379.36
Synonym 4-[(RS)-(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenyl Sodium Sulphate, Sodium Picosulfate Imp. A (EP), 4-[(Pyridin-2-yl)(4-hydroxyphenyl)methyl]phenyl Sodium Sulphate
IUPAC Name sodium;[4-[(4-hydroxyphenyl)-pyridin-2-ylmethyl]phenyl] sulfate
SMILES [Na+].Oc1ccc(cc1)C(c2ccc(OS(=O)(=O)[O-])cc2)c3ccccn3
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C18H15NO5S.Na/c20-15-8-4-13(5-9-15)18(17-3-1-2-12-19-17)14-6-10-16(11-7-14)24-25(21,22)23;/h1-12,18,20H,(H,21,22,23);/q;+1/p-1
Formula Weight 379.049
Percent Purity >95%
Chemical Name or Material 4-[(RS)-(4-Hydroxyphenyl)(pyridin-2-yl)methyl]phenyl Sodium Sulfate (4-[(Pyridin-2-yl)(4-hydroxyphenyl)methyl]phenyl Sodium Sulfate)
Grade HPLC