Learn More
4-Cyanopyrazole, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Cyanopyrazole |
| CAS | 31108-57-3 |
| Molecular Formula | C4H3N3 |
| Quantity | 1 g |
| Synonym | Pyrazole-4-carbonitrile, 1H-Pyrazole-4-carbonitrile |
| InChI Formula | InChI=1S/C4H3N3/c5-1-4-2-6-7-3-4/h2-3H,(H,6,7) |
| SMILES | N#Cc1cn[nH]c1 |
| IUPAC Name | 1H-pyrazole-4-carbonitrile |
| Molecular Weight (g/mol) | 93.09 |
| Formula Weight | 93.0327 |